SY3H3D678844 Synthonix Corporation

5-(4-Bromophenyl)-6-hydroxypyrimidin-4(1H)-one

  • MDL number MFCD28044068
  • Cas Number : 706811-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53526.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/53526.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart