SY3H3D67887B Synthonix Corporation

5-Bromo-1-methyl-1H-indole-3-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 400071-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53751.html

Safety Information

MSDS https://synthonix.com/53751.html
Availability details for
Quantity
  • Availability
  • Add to Cart