SY3H3D67888A Synthonix Corporation

1-(4-Bromo-2-fluorophenyl)cyclopropanecarboxylic acid

  • MDL number MFCD12405622
  • Cas Number : 872422-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53799.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53799.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart