SY3H3D67888E Synthonix Corporation

5-Bromo-7-fluoro-3,4-dihydronaphthalen-1(2H)-one

  • MDL number MFCD18208383
  • Cas Number : 1260018-37-8
  • Molecular Weight : 243.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/53827.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/53827.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart