SY3H3D6788AE Synthonix Corporation

2-Bromo-3,4-dihydronaphthalen-1(2H)-one

  • MDL number MFCD00089999
  • Cas Number : 13672-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54045.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54045.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart