SY3H3D6788B0 Synthonix Corporation

4-Bromo-2-methyl-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD09881709
  • Cas Number : 174702-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Lump / Colour: White - Pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54047.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54047.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart