SY3H3D6788C1 Synthonix Corporation

tert-Butyl 3-(aminomethyl)morpholine-4-carboxylate hydrochloride

  • MDL number MFCD27920927
  • Cas Number : 1802489-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54224.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54224.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart