SY3H3D6788D3 Synthonix Corporation

2-(((Benzyloxy)carbonyl)amino)-3,3,3-trifluoropropanoic acid

  • MDL number MFCD10697679
  • Cas Number : 10068-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54337.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54337.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart