SY3H3D6788D7 Synthonix Corporation

1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD11845717
  • Cas Number : 294877-29-5
  • Molecular Weight : 251.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Light-yellow to brown powder or crystals or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54378.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54378.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart