SY3H3D6788F5 Synthonix Corporation

3-(Benzoylthio)-2-methylpropanoic acid

  • MDL number MFCD09864827
  • Cas Number : 74431-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54584.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54584.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart