SY3H3D6788F7 Synthonix Corporation

(S)-2-Benzylsuccinic acid

  • MDL number MFCD00798341
  • Cas Number : 3972-36-9
  • Molecular Weight : 208.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54586.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54586.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart