SY3H3D6788FD Synthonix Corporation

6-Bromo-2,3,4,4a,5,9b-hexahydro-1H-pyrido[4,3-b]indole

  • MDL number MFCD29923356
  • Cas Number : 1059630-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54606.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54606.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart