SY3H3D67890A Synthonix Corporation

5-Bromo-3-(tert-butyl)-2-hydroxybenzaldehyde

  • MDL number MFCD00456060
  • Cas Number : 153759-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54674.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54674.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart