SY3H3D67890D Synthonix Corporation

4-(tert-Butyl)-3-iodobenzoic acid

  • MDL number MFCD11110767
  • Cas Number : 91131-72-5
  • Molecular Weight : 304.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54681.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54681.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart