SY3H3D67891F Synthonix Corporation

(4-Bromophenyl)(cyclobutyl)methanamine hydrochloride

  • MDL number MFCD12912905
  • Cas Number : 1193387-95-9
  • Molecular Weight : 276.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54729.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54729.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart