SY3H3D678930 Synthonix Corporation

3-((Benzyloxy)methyl)oxetane

  • MDL number MFCD20482348
  • Cas Number : 1003013-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54769.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54769.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart