SY3H3D678936 Synthonix Corporation

1-Bromo-2,3,4,5,6-pentamethylbenzene

  • MDL number MFCD00013523
  • Cas Number : 5153-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54817.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54817.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart