SY3H3D67893E Synthonix Corporation

tert-Butyl ((6-chloropyridin-2-yl)methyl)carbamate

  • MDL number MFCD27923671
  • Cas Number : 1640995-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54832.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54832.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart