SY3H3D67894D Synthonix Corporation

N,N-Bis(2-chloroethyl)-4-methylbenzenesulfonamide

  • MDL number MFCD00018944
  • Cas Number : 42137-88-2
  • Molecular Weight : 296.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54885.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54885.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart