SY3H3D678D61 Synthonix Corporation

tert-Butyl 5-methyl-2,4-dioxopiperidine-1-carboxylate

  • MDL number MFCD24466520
  • Cas Number : 942425-69-6
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54999.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54999.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart