SY3H3D678D77 Synthonix Corporation

3-((tert-Butoxycarbonyl)amino)-3-cyclopropylpropanoic acid

  • MDL number MFCD01861819
  • Cas Number : 683218-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55052.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55052.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart