SY3H3D678D90 Synthonix Corporation

Butyl Butyryllactate

  • MDL number MFCD00027213
  • Cas Number : 7492-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55091.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55091.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart