SY3H3D678D99 Synthonix Corporation

(S)-3-((tert-Butoxycarbonyl)amino)-4-methylpentanoic acid

  • MDL number MFCD01076234
  • Cas Number : 179412-79-4
  • Molecular Weight : 231.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form powder Colour: white to pale-yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55105.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55105.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart