SY3H3D678D9C Synthonix Corporation

tert-butyl (cis-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)carbamate

  • MDL number MFCD23159250
  • Cas Number : 1923238-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55109.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55109.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart