SY3H3D678D9E Synthonix Corporation

(R)-2-((tert-Butoxycarbonyl)amino)-4-methylpentanoic acid hydrate

  • MDL number MFCD00065583
  • Cas Number : 200937-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55111.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55111.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart