SY3H3D678DCB Synthonix Corporation

8-Bromo-1-naphthoic acid

  • MDL number No MDL found
  • Cas Number : 1729-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55202.html

Safety Information

MSDS https://synthonix.com/55202.html
Availability details for
Quantity
  • Availability
  • Add to Cart