SY3H3D678DE4 Synthonix Corporation

3-(4-Bromophenyl)pyridine

  • MDL number MFCD04116232
  • Cas Number : 129013-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55257.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55257.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart