SY3H3D678DEC Synthonix Corporation

2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid

  • MDL number MFCD00221424
  • Cas Number : 35480-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Very pale yellow - Pale-red
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55300.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55300.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart