SY3H3D678DF9 Synthonix Corporation

6-Bromo-2-naphthylmethanol

  • MDL number MFCD09842470
  • Cas Number : 100751-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55418.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/55418.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart