SY3H3D678E08 Synthonix Corporation

tert-Butyl (6-methylpiperidin-3-yl)carbamate hydrochloride

  • MDL number MFCD28401030
  • Cas Number : 1807542-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55543.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55543.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart