SY3H3D678E20 Synthonix Corporation

3-(Benzyloxy)-5-bromopyridin-2-amine

  • MDL number MFCD11109946
  • Cas Number : 754230-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Pale-yellow to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55645.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55645.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart