SY3H3D678E23 Synthonix Corporation

2-(Benzylthio)-9H-purin-6-amine

  • MDL number MFCD30729982
  • Cas Number : 64542-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Off-white to light yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55653.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/55653.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart