SY3H3D678E2C Synthonix Corporation

6-Bromo-2,3,4,9-tetrahydro-1H-carbazole

  • MDL number MFCD00094989
  • Cas Number : 21865-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55745.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55745.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart