SY3H3D678E3C Synthonix Corporation

1-Benzoylpiperidin-4-one

  • MDL number MFCD00006189
  • Cas Number : 24686-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Very pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55809.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55809.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart