SY3H3D678E3E Synthonix Corporation

(S)-4-Benzyl-5-oxomorpholine-3-carboxylic acid

  • MDL number MFCD09835535
  • Cas Number : 106973-37-9
  • Molecular Weight : 235.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55835.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55835.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart