SY3H3D678E44 Synthonix Corporation

7-Bromo-5,8-dimethyl-3,4-dihydronaphthalen-1(2H)-one

  • MDL number MFCD19671389
  • Cas Number : 30098-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55851.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55851.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart