SY3H3D678E4F Synthonix Corporation

9-Bromo-1,2,3,5,6,7-hexahydropyrido[3,2,1-ij]quinoline

  • MDL number No MDL found
  • Cas Number : 70173-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55894.html

Safety Information

MSDS https://synthonix.com/55894.html
Availability details for
Quantity
  • Availability
  • Add to Cart