SY3H3D678E6B Synthonix Corporation

2-Benzamido-3-methylbutanoic acid

  • MDL number MFCD00038283
  • Cas Number : 2901-80-6
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55987.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55987.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart