SY3H3D678E73 Synthonix Corporation

1-Benzylpiperidin-4-one oxime

  • MDL number MFCD00277794
  • Cas Number : 949-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56022.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56022.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart