SY3H3D678E79 Synthonix Corporation

3-((tert-Butoxycarbonyl)amino)-2-methoxyisonicotinic acid

  • MDL number MFCD22199249
  • Cas Number : 870997-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56043.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56043.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart