SY3H3D678E88 Synthonix Corporation

(S)-4-Benzyl-3-(chloromethyl)morpholine

  • MDL number MFCD10687133
  • Cas Number : 917572-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Colorless Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56066.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56066.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart