SY3H3D678E94 Synthonix Corporation

tert-Butyl (2-bromo-4,5,6,7-tetrahydrobenzo[d]thiazol-6-yl)carbamate

  • MDL number MFCD13195380
  • Cas Number : 1251009-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56118.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56118.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart