SY3H3D678E9A Synthonix Corporation

(1R,2R)-2-(Benzylamino)cyclopentanol

  • MDL number MFCD09999250
  • Cas Number : 1033605-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56139.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56139.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart