SY3H3D678E9E Synthonix Corporation

(R)-(4-Benzylmorpholin-3-yl)methanol

  • MDL number No MDL found
  • Cas Number : 101376-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56146.html

Safety Information

MSDS https://synthonix.com/56146.html
Availability details for
Quantity
  • Availability
  • Add to Cart