SY3H3D678EA3 Synthonix Corporation

tert-Butyl 6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate

  • MDL number MFCD11518948
  • Cas Number : 230301-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56168.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56168.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart