SY3H3D678EB3 Synthonix Corporation

tert-Butyl 4-cyano-2,2-dimethyl-3-oxopyrrolidine-1-carboxylate

  • MDL number MFCD08275049
  • Cas Number : 946497-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56204.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56204.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart