SY3H3D678EC2 Synthonix Corporation

tert-Butyl 3-((dimethylamino)methylene)-4-oxopyrrolidine-1-carboxylate

  • MDL number MFCD11616027
  • Cas Number : 157327-42-9
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/56256.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56256.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart