SY3H3D678EC3 Synthonix Corporation

tert-Butyl 2-((tert-butoxycarbonyl)amino)-2-cyanoacetate

  • MDL number MFCD24466150
  • Cas Number : 130892-40-9
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56258.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56258.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart