SY3H3D678EC6 Synthonix Corporation

tert-Butyl ((5-carbamoyl-1-methyl-1H-pyrazol-3-yl)methyl)(methyl)carbamate

  • MDL number MFCD30566064
  • Cas Number : 1643141-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56265.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56265.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart