SY3H3D678EC7 Synthonix Corporation

1-N-Boc-2-Ethylpiperidin-4-one

  • MDL number MFCD07367729
  • Cas Number : 324769-07-5
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/56274.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/56274.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart